Tryptic Soy Agar (TSA), CRITERION™, Hardy Diagnostics
Fournisseur: HARDY DIAGNOSTICS CA
Certificats
À propos de cet article
CRITERION™ Tryptic Soy Agar is recommended for use as a general growth medium for the isolation and cultivation of microorganisms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
This medium is also recommended for use in the cultivation, storage, and transportation of pure cultures of bacteria.
Certifications: BSE Free
Spécifications
- Storage temperature:–2-30°C