Rappaport-Vassiliadis Broth, Hardy Diagnostics
Hardy Diagnostics' CRITERION™ Rappaport-Vassiliadis Broth is recommended for the selective enrichment of Salmonella spp. from food samples.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
CRITERION™ Dehydrated Culture Media by Hardy Diagnostics is formulated to meet or exceed the highest standards. Compare these features…
Hardy Diagnostics' CRITERION™ Rappaport-Vassiliadis Broth is recommended for the selective enrichment of Salmonella spp
from food samples.
: BSE Free
- Storage temperature:–2-30°C
Frequently Bought Together