MacConkey Agar, CRITERION™, Hardy Diagnostics
About this item
CRITERION™ MacConkey Agar is recommended for use as a selective and differential medium for the isolation of gram-negative bacilli (including coliform organisms and enteric pathogens), on the basis of lactose fermentation
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
4 Options Available Below
Product Details & Documents
CRITERION™ MacConkey Agar is recommended for use as a selective and differential medium for the isolation of gram-negative bacilli (including coliform organisms and enteric pathogens), on the basis of lactose fermentation
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- CA89405-628
- CA89405-630
- CA89405-632
- CA89405-634
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Specifications
Product Family Options
Product Information
- TailleFinal VolumeType de conditionnementAvailabilityPrice
- Supplier Number: C6130Specifications:
More
Specifications
Cat. No.CA89405-628Storage temperature–2-30°C - Supplier Number: C6131Specifications:
More
Specifications
Cat. No.CA89405-630Storage temperature–2-30°C - Supplier Number: C6132Specifications:
More
Specifications
Cat. No.CA89405-632Storage temperature–2-30°C - Supplier Number: C6133Specifications:
More
Specifications
Cat. No.CA89405-634Storage temperature–2-30°C