MacConkey Broth, CRITERION™, Hardy Diagnostics
About this item
CRITERION™ MacConkey Broth is recommended for cultivating gram-negative, lactose-fermenting bacilli from water and foods as a presumptive test for coliforms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
MacConkey Broth can also be used preenrichment of E. coli O157 for toxin testing. Dehydratedculture media with engineered container.
Certifications: BSE Free
4 Options Available Below
Product Details & Documents
CRITERION™ MacConkey Broth is recommended for cultivating gram-negative, lactose-fermenting bacilli from water and foods as a presumptive test for coliforms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
MacConkey Broth can also be used preenrichment of E. coli O157 for toxin testing. Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- CA89405-636
- CA89405-638
- CA89405-640
- CA89405-642
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Specifications
Product Family Options
Product Information
- TailleFinal VolumeType de conditionnementAvailabilityPrice
- Supplier Number: C6140Specifications:
More
Specifications
Cat. No.CA89405-636Storage temperature–2-30°C - Supplier Number: C6141Specifications:
More
Specifications
Cat. No.CA89405-638Storage temperature–2-30°C - Supplier Number: C6142Specifications:
More
Specifications
Cat. No.CA89405-640Storage temperature–2-30°C - Supplier Number: C6143Specifications:
More
Specifications
Cat. No.CA89405-642Storage temperature–2-30°C