m FC Broth Base, CRITERION™, Hardy Diagnostics
Fournisseur: HARDY DIAGNOSTICS CA
Certificats
À propos de cet article
CRITERION™ m FC Broth Base is recommended for use with added Rosolic Acid for the cultivation and enumeration of fecal coliforms using membrane filtration technique at elevated temperatures
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Spécifications
- Storage temperature:–2-30°C