EC Medium, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
About this item
CRITERION™ EC Medium is recommended for the detection of coliforms at 35 degrees C
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
and Escherichia coli and fecal coliforms at 45.5 degrees C. Dehydratedculture media with engineered container.
Certifications: BSE Free
2 Options Available Below
Product Details & Documents
CRITERION™ EC Medium is recommended for the detection of coliforms at 35 degrees C
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
and Escherichia coli and fecal coliforms at 45.5 degrees C. Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- CA89405-422
- CA89405-424
- Storage temperature:
- –2-30°C
- –2-30°C
Specifications
Product Family Options
Product Information
- TailleType de conditionnementAvailabilityPrice
- Supplier Number: C5691Specifications:
More
Specifications
Cat. No.CA89405-422Storage temperature–2-30°C - Supplier Number: C5692Specifications:
More
Specifications
Cat. No.CA89405-424Storage temperature–2-30°C