Deoxycholate Lactose Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
À propos de cet article
CRITERION™ Deoxycholate Lactose Agar for cultivation, maintenance, fermentation reaction and motility determination of bacteria
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
3 Options Available Below
Product Details & Documents
CRITERION™ Deoxycholate Lactose Agar for cultivation, maintenance, fermentation reaction and motility determination of bacteria
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- CA89405-356
- CA89405-358
- CA89405-360
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
Spécifications
Product Family Options
Informations sur le produit
- TailleType de conditionnementAvailabilityPrice
- Supplier Number: C5561Spécifications:
More
Specifications
Cat. No.CA89405-356Storage temperature–2-30°C - Supplier Number: C5562Spécifications:
More
Specifications
Cat. No.CA89405-358Storage temperature–2-30°C - Supplier Number: C5563Spécifications:
More
Specifications
Cat. No.CA89405-360Storage temperature–2-30°C