TAT Broth | Avantor
TAT Broth
TAT Broth
Certificates
TAT Broth
5 Options Available Below
Product Details & Documents
For the cultivation of microorganisms from highly viscous or gelatinous materials.
  • Microorganism cultivation
  • Diagnosis of human disease
  • Contains preservatives for self-sterilization against vegetative bacteria, yeast, and mold
Hardy Diagnostics TAT (Tryptone-Azolectin-Tween®) Broth is recommended for use in cultivating microorganisms from highly viscous or gelatinous materials, such as salves and ointments.
This product is not intended to be used for the diagnosis of human disease.
TAT (Tryptone-Azolectin-Tween) Broth, also known as Casein-peptone Lecithin Polysorbate Broth, is recommended for sterility testing of viscous materials, such as salves, creams and ointments, and is especially suited to the sterility testing of cosmetics. Cosmetics and topical pharmaceutical products are easily subjected to contamination during the manufacturing process and through continued use by consumers. Consequently, these products contain preservatives for self-sterilization against vegetative bacteria, yeast, and mold.
Hardy Diagnostics TAT Broth formulations are enriched to isolate and cultivate microorganisms from viscous materials. These media contain pancreatic digest of casein, which provides nitrogen, vitamins, amino acids, and carbon. Lecithin (azolectin) and Tween® 20 neutralize common preservatives in cosmetics or pharmaceutical products, thereby permitting the growth of contaminating flora. The relatively high casein content provides optimal conditions for spore germination and regeneration of damaged cells. Lecithin and polysorbate 20 inactivate many antimicrobial compounds.
These media are especially adapted to the microbiological examination of non-sterile products from pharmaceutical, cosmetic and raw material or end-products, for the purpose of performing microbial enumeration studies.
Product Family Options
Informations sur le produit
  • Taille
    Type de conditionnement
    Température de stockage
    Availability
    Price
  • 9 ml
    Tube
    2...30 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 20
    Pack of 20 - $108.00
    $108.00
    Pack of 20
       $108.00
    La quantité minimale de commande est de 1
    Supplier Number: K251
  • 90 ml
    Glass jar, wide mouth
    2...30 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $201.00
    $201.00
    Pack of 12
       $201.00
    La quantité minimale de commande est de 1
    Supplier Number: U77
  • 90 ml
    Bottle, Boston round
    2...30 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 20
    Pack of 20 - $321.00
    $321.00
    Pack of 20
       $321.00
    La quantité minimale de commande est de 1
    Supplier Number: U87
  • 99 ml
    Bottle, Boston round
    2...30 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 20
    Pack of 20 - $357.00
    $357.00
    Pack of 20
       $357.00
    La quantité minimale de commande est de 1
    Supplier Number: U78
  • 1 L
    PC bottle
    2...30 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $783.00
    $783.00
    Pack of 10
       $783.00
    La quantité minimale de commande est de 1
    Supplier Number: U319