Rappaport-Vassiliadis Broth, USP, Hardy Diagnostics
Catalog # CA89407-854
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Pack type:Tube
- Size:10 mL
- Storage temperature:2-8°C
- Cat. no.:CA89407-854
- Supplier No.:K246
Specifications
About this item
Recommended for the selective enrichment of Salmonella species from food samples, and conforms to the Harmonized USP/EP/JP requirements.