Lauryl Tryptose Broth, Hardy Diagnostics
For the detection of coliforms in water and food
Contains a durham tube for gas detection.
For the detection of coliforms in water and food
Contains a durham tube for gas detection.
- Catalog No:
- 75813-966
- CA89407-684
- CA89407-686
- CA89407-688
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
Product Family Options
- DésignationTailleType de conditionnementDimensions
- Double strength10 mLTube, Durham20×125 mmPack of 20 (10ml)$165.00: K338Product available on GSA Advantage®
Specifications
Cat. No.75813-966Storage temperature - : K33
Specifications
Cat. No.CA89407-684Storage temperature–2-30°C - : K32
Specifications
Cat. No.CA89407-686Storage temperature–2-30°C - : K61
Specifications
Cat. No.CA89407-688Storage temperature–2-30°C