Fluid Thioglycollate Broth, Hardy Diagnostics
Fournisseur: HARDY DIAGNOSTICS CA
Certificats
À propos de cet article
For the cultivation of microorganisms and USP sterility procedures
Filtered to remove all dead bacteria and debris.
Spécifications
- Storage conditions:–2-30°C