EE Broth Mossel
EE Broth Mossel
Certificates
EE Broth Mossel
2 Options Available Below
Product Details & Documents
For use in the selective enrichment, isolation, and enumeration of Enterobacteriaceae, particularly from foods. Conforms with harmonized USP/EP/JP requirements.
  • Contains glucose to facilitate the growth of most Enterobacteriaceae
  • EE Broth Mossel, USP should be used as an enrichment broth
  • Contains peptones as a source of nitrogen, vitamins and amino acids
EE Broth Mossel, USP conforms to the formulation as specified in the U.S
The detection of Enterobacteriaceae is of great concern in the microbial examination of non-sterile products and in monitoring the sanitary quality of foods. Enterobacteriaceae are easily injured during processing procedures, exposure to low temperatures, sub-marginal heat, drying, radiation, and preservatives or sanitizers. Therefore, the ability to recover these microorganisms depends upon proper resuscitation of sub-lethally damaged cells.
EE Broth Mossel, USP contains peptones as a source of nitrogen, vitamins and amino acids. Dextrose provides a carbon source. Phosphates act as buffering agents, while brilliant green and oxbile (oxgall) are added as selective agents.
Pharmacopoeia (USP), which is based on the formula proposed by Mossel, Visser and Cornelissen. The formula contains glucose to facilitate the growth of most Enterobacteriaceae and aids in the detection of Salmonella and other lactose-negative organisms. EE Broth Mossel, USP should be used as an enrichment broth, followed by subculture to a selective solid medium (e.g. Violet Red Bile with Glucose Agar).
Specifications for Product Family
Spécifications
Spécifications
  • Catalog No:
  • CA89407-508
  • CA89407-506
  • Storage temperature:
  • –2...+30 °C
  • –2...+30 °C
Spécifications
Product Family Options
Informations sur le produit
  • Taille
    Type de conditionnement
    Availability
    Price
  • 9 ml
    Glass tube, 16×125mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 20
    Pack of 20 - $320.00
    $320.00
    Pack of 20
       $320.00
    Supplier Number: K191
    Spécifications:
    More
    Specifications
    Cat. No.
    CA89407-508
    Storage temperature
    –2...+30 °C
  • 100 ml
    PC jar
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $359.00
    $359.00
    Pack of 12
       $359.00
    Supplier Number: U391
    Spécifications:
    More
    Specifications
    Cat. No.
    CA89407-506
    Storage temperature
    –2...+30 °C