ContactUsLinkComponent
CT-SMAC with Cefi x ime and Tellurite, Hardy Diagnostics
Catalogue # CA89407-158
Fournisseur:  HARDY DIAGNOSTICS CA
Numéro CAS:  
undefined
CT-SMAC with Cefi x ime and Tellurite, Hardy Diagnostics
Catalogue # CA89407-158
Fournisseur:  HARDY DIAGNOSTICS CA
Supplier Number:  G129
Numéro CAS:  
Spécifications
  • Pack type:
    Plate
  • Size:
    15 x 100 mm
  • Storage temperature:
    2-8°C
  • Cat. no.:
    CA89407-158
  • Volume:
    18 ml
  • Supplier No.:
    G129
Spécifications
À propos de cet article
For selection and differentiation of enterohemorrhagic E.coli O157.