Butterfield's Phosphate Buffer, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
À propos de cet article
For the preparation of dilutions for plate count and other laboratory tests, pH 7.2.
7 Options Available Below
Product Details & Documents
For the preparation of dilutions for plate count and other laboratory tests, pH 7.2.
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- CA89407-406
- CA89407-408
- CA89407-410
- CA89407-412
- CA89407-414
- CA89407-416
- CA89407-440
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Spécifications
Product Family Options
Informations sur le produit
- DésignationTailleType de conditionnementAvailabilityPrice
- Supplier Number: U150Spécifications:
More
Specifications
Cat. No.CA89407-406Storage temperature–2-30°C - Supplier Number: K109Spécifications:
More
Specifications
Cat. No.CA89407-408Storage temperature–2-30°C - Supplier Number: K208Spécifications:
More
Specifications
Cat. No.CA89407-410Storage temperature–2-30°C - Supplier Number: K119Spécifications:
More
Specifications
Cat. No.CA89407-412Storage temperature–2-30°C - Supplier Number: U190Spécifications:
More
Specifications
Cat. No.CA89407-414Storage temperature–2-30°C - Supplier Number: K209Spécifications:
More
Specifications
Cat. No.CA89407-416Storage temperature–2-30°C - Supplier Number: U290Spécifications:
More
Specifications
Cat. No.CA89407-440Storage temperature–2-30°C