Urea Base Concentrate, CRITERION™, Hardy Diagnostics
Fournisseur: HARDY DIAGNOSTICS CA
Certificats
À propos de cet article
CRITERION™ Urea Base Concentrate is a 10X concentrate for the differentiation of microorganisms based on urease production.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Certifications: BSE Free
Spécifications
- Storage temperature:–2-30°C