Thioglycollate Medium without Indicator, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Thioglycollate Medium without Indicator, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Certificates
À propos de cet article
CRITERION™ Thioglycollate Medium without Indicator is recommended for the cultivation of aerobic, microaerophilic, and anaerobic microorganisms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
4 Options Available Below
Product Details & Documents
CRITERION™ Thioglycollate Medium without Indicator is recommended for the cultivation of aerobic, microaerophilic, and anaerobic microorganisms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- CA89406-134
- CA89406-136
- CA89406-138
- CA89406-140
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Spécifications
Product Family Options
Informations sur le produit
- TailleFinal VolumeType de conditionnementAvailabilityPrice
- Supplier Number: C7080Spécifications:
More
Specifications
Cat. No.CA89406-134Storage temperature–2-30°C - Supplier Number: C7081Spécifications:
More
Specifications
Cat. No.CA89406-136Storage temperature–2-30°C - Supplier Number: C7082Spécifications:
More
Specifications
Cat. No.CA89406-138Storage temperature–2-30°C - Supplier Number: C7083Spécifications:
More
Specifications
Cat. No.CA89406-140Storage temperature–2-30°C