CRITERION™ Sucrose
À propos de cet article
CRITERION™ Sucrose is added to culture media for use in preparing microbiological culture media.
- Wide mouth opening
- Reduces dust and allows easy scooping
- Convenient handgrip
- 'No slip' finger holds
- Stackable containers
- High solubility; dissolves quickly
- Custom and bulk products available
- Unparalleled performance - tested for optimal growth
For fermentation tests it is usually added at a final concentration of 0.5 to 1.0%. Dehydratedculture media with engineered container.
These culture media is formulated to meet or exceed the highest standards.
Application: Breweries, wineries and distilleries, cannabis QA testing, clinical, cosmetics, environmental, food safety, compounding pharmacy, pharmaceutical manufacturing
Certifications: TSE/BSE free.
Delivery information: 500 g - wide mouth, easy grip bottle, comes with a free scoop, 2 and 10 kg bucket - with convenient screw lid.
4 Options Available Below
Product Details & Documents
CRITERION™ Sucrose is added to culture media for use in preparing microbiological culture media.
- Wide mouth opening
- Reduces dust and allows easy scooping
- Convenient handgrip
- 'No slip' finger holds
- Stackable containers
- High solubility; dissolves quickly
- Custom and bulk products available
- Unparalleled performance - tested for optimal growth
For fermentation tests it is usually added at a final concentration of 0.5 to 1.0%. Dehydratedculture media with engineered container.
These culture media is formulated to meet or exceed the highest standards.
Application: Breweries, wineries and distilleries, cannabis QA testing, clinical, cosmetics, environmental, food safety, compounding pharmacy, pharmaceutical manufacturing
Certifications: TSE/BSE free.
Delivery information: 500 g - wide mouth, easy grip bottle, comes with a free scoop, 2 and 10 kg bucket - with convenient screw lid.
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- CA89406-094
- CA89406-096
- CA89406-098
- CA89406-100
- Storage temperature:
- 2...30 °C
- 2...30 °C
- 2...30 °C
- 2...30 °C
Spécifications
Product Family Options
Informations sur le produit
- TailleType de conditionnementAvailabilityPrice
- Supplier Number: C7020Spécifications:
More
Specifications
Cat. No.CA89406-094Storage temperature2...30 °C - Supplier Number: C7021Spécifications:
More
Specifications
Cat. No.CA89406-096Storage temperature2...30 °C - Supplier Number: C7022Spécifications:
More
Specifications
Cat. No.CA89406-098Storage temperature2...30 °C - Supplier Number: C7023Spécifications:
More
Specifications
Cat. No.CA89406-100Storage temperature2...30 °C