Staphylococcus 110 Agar, CRITERION™, Hardy Diagnostics | Avantor
Staphylococcus 110 Agar, CRITERION™, Hardy Diagnostics
Staphylococcus 110 Agar, CRITERION™, Hardy Diagnostics
Certificates
Staphylococcus 110 Agar, CRITERION™, Hardy Diagnostics
Staphylococcus 110 Agar, CRITERION™, Hardy Diagnostics
2 Options Available Below
Product Details & Documents
CRITERION™ Staphylococcus Medium 110 is used for isolating and differentiating pathogenic staphylococci based on fermentation, pigment formation and gelatinase activity
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications for Product Family
Spécifications
Spécifications
  • Catalog No:
  • CA89406-070
  • CA89406-072
  • Storage temperature:
  • –2-30°C
  • –2-30°C
Spécifications
Product Family Options
Informations sur le produit
  • Taille
    Final Volume
    Type de conditionnement
    Availability
    Price
  • 122.4 g
    2 L
    Zip-pouch to make 2 L, Mylar®
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $531.00
    $531.00
    Each
       $531.00
    La quantité minimale de commande est de 1
    Supplier Number: C6990
    Spécifications:
    More
    Specifications
    Cat. No.
    CA89406-070
    Storage temperature
    –2-30°C
  • 500 g
    Bottle, wide mouth
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $752.00
    $752.00
    Each
       $752.00
    La quantité minimale de commande est de 1
    Supplier Number: C6991
    Spécifications:
    More
    Specifications
    Cat. No.
    CA89406-072
    Storage temperature
    –2-30°C