Standard Methods Agar, CRITERION™, Hardy Diagnostics
About this item
CRITERION™ Standard Methods Agar (Plate Count Agar) is recommended for use in determining the microbial content in dairy products, food, water and other material of sanitary importance
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
4 Options Available Below
Product Details & Documents
CRITERION™ Standard Methods Agar (Plate Count Agar) is recommended for use in determining the microbial content in dairy products, food, water and other material of sanitary importance
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- CA89406-062
- CA89406-064
- CA89406-066
- CA89406-068
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Specifications
Product Family Options
Product Information
- TailleFinal VolumeType de conditionnementAvailabilityPrice
- Supplier Number: C6980Specifications:
More
Specifications
Cat. No.CA89406-062Storage temperature–2-30°C - Supplier Number: C6981Specifications:
More
Specifications
Cat. No.CA89406-064Storage temperature–2-30°C - Supplier Number: C6982Specifications:
More
Specifications
Cat. No.CA89406-066Storage temperature–2-30°C - Supplier Number: C6983Specifications:
More
Specifications
Cat. No.CA89406-068Storage temperature–2-30°C