SIM Medium, CRITERION™, Hardy Diagnostics
À propos de cet article
CRITERION™ SIM Medium is recommended for the differentiation of gram-negative enteric bacilli, such as Salmonella and Shigella, on the basis of sulfide production, indole formation, and motility
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
4 Options Available Below
Product Details & Documents
CRITERION™ SIM Medium is recommended for the differentiation of gram-negative enteric bacilli, such as Salmonella and Shigella, on the basis of sulfide production, indole formation, and motility
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- CA89406-038
- CA89406-040
- CA89406-042
- CA89406-044
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Spécifications
Product Family Options
Informations sur le produit
- TailleFinal VolumeType de conditionnementAvailabilityPrice
- Supplier Number: C6940Spécifications:
More
Specifications
Cat. No.CA89406-038Storage temperature–2-30°C - Supplier Number: C6941Spécifications:
More
Specifications
Cat. No.CA89406-040Storage temperature–2-30°C - Supplier Number: C6942Spécifications:
More
Specifications
Cat. No.CA89406-042Storage temperature–2-30°C - Supplier Number: C6943Spécifications:
More
Specifications
Cat. No.CA89406-044Storage temperature–2-30°C