CRITERION™ Purple Broth Base | Avantor
CRITERION™ Purple Broth Base
CRITERION™ Purple Broth Base
Certificates
CRITERION™ Purple Broth Base
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
CRITERION™ purple broth base is primarily for use with added carbohydrate in differentiating pure cultures of bacteria, particularly of enteric organisms, based on fermentation reactions.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
  • Unparalleled performance - tested for optimal growth
Dehydrated culture media with engineered container.
These culture media is formulated to meet or exceed the highest standards.
Application: Breweries, wineries and distilleries, cannabis QA testing, clinical, cosmetics, environmental, food safety, compounding pharmacy, pharmaceutical manufacturing.
Certifications: BSE free.
Delivery information: 500 g - wide mouth, easy grip bottle, comes with a free scoop, 2 and 10 kg bucket - with convenient screw lid.
Specifications for Product Family
Spécifications
Spécifications
  • Catalog No:
  • CA89405-944
  • CA89405-946
  • Storage temperature:
  • 2...30 °C
  • 2...30 °C
Spécifications
Product Family Options
Informations sur le produit
  • Taille
    Type de conditionnement
    Availability
    Price
  • 122.4 g
    Zip-pouch to make 2 L, Mylar®
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 116,00 $
    116,00 $
    Each
       116,00 $
    Supplier Number: C6710
    Spécifications:
    More
    Specifications
    Cat. No.
    CA89405-944
    Storage temperature
    2...30 °C
  • 500 g
    Bottle, wide mouth
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 393,00 $
    393,00 $
    Each
       393,00 $
    La quantité minimale de commande est de 1
    Supplier Number: C6711
    Spécifications:
    More
    Specifications
    Cat. No.
    CA89405-946
    Storage temperature
    2...30 °C