PC Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
À propos de cet article
CRITERION™ PC (Pseudomonas cepacia) Agar is recommended for the selective isolation of Burkholderia (Pseudomonas) cepacia from clinical and environmental materials
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
4 Options Available Below
Product Details & Documents
CRITERION™ PC (Pseudomonas cepacia) Agar is recommended for the selective isolation of Burkholderia (Pseudomonas) cepacia from clinical and environmental materials
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- CA89405-928
- CA89405-930
- CA89405-932
- CA89405-934
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Spécifications
Product Family Options
Informations sur le produit
- TailleFinal VolumeType de conditionnementAvailabilityPrice
- Supplier Number: C6660Spécifications:
More
Specifications
Cat. No.CA89405-928Storage temperature–2-30°C - Supplier Number: C6661Spécifications:
More
Specifications
Cat. No.CA89405-930Storage temperature–2-30°C - Supplier Number: C6662Spécifications:
More
Specifications
Cat. No.CA89405-932Storage temperature–2-30°C - Supplier Number: C6663Spécifications:
More
Specifications
Cat. No.CA89405-934Storage temperature–2-30°C