Nutrient Broth, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
À propos de cet article
CRITERION™ Nutrient Broth is a general purpose growth media recommended for use in the cultivation of nonfastidious microorganisms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container. Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience.
Certifications: BSE Free
4 Options Available Below
Product Details & Documents
CRITERION™ Nutrient Broth is a general purpose growth media recommended for use in the cultivation of nonfastidious microorganisms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container. Custom and bulk products available. BSE Free documentation available. Over 30 years of formulation experience.
Certifications: BSE Free
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- CA89405-792
- CA89405-794
- CA89405-796
- CA89405-798
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Spécifications
Product Family Options
Informations sur le produit
- TailleType de conditionnementAvailabilityPrice
- Supplier Number: C6470Spécifications:
More
Specifications
Cat. No.CA89405-792Storage temperature–2-30°C - Supplier Number: C6471Spécifications:
More
Specifications
Cat. No.CA89405-794Storage temperature–2-30°C - Supplier Number: C6472Spécifications:
More
Specifications
Cat. No.CA89405-796Storage temperature–2-30°C - Supplier Number: C6473Spécifications:
More
Specifications
Cat. No.CA89405-798Storage temperature–2-30°C