Nutrient Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
À propos de cet article
CRITERION™ Nutrient Agar is a general purpose growth media recommended for use in the isolation and cultivation of nonfastidious microorganisms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
4 Options Available Below
Product Details & Documents
CRITERION™ Nutrient Agar is a general purpose growth media recommended for use in the isolation and cultivation of nonfastidious microorganisms
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- CA89405-784
- CA89405-786
- CA89405-788
- CA89405-790
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Spécifications
Product Family Options
Informations sur le produit
- TailleFinal VolumeType de conditionnementAvailabilityPrice
- Supplier Number: C6460Spécifications:
More
Specifications
Cat. No.CA89405-784Storage temperature–2-30°C - Supplier Number: C6461Spécifications:
More
Specifications
Cat. No.CA89405-786Storage temperature–2-30°C - Supplier Number: C6462Spécifications:
More
Specifications
Cat. No.CA89405-788Storage temperature–2-30°C - Supplier Number: C6463Spécifications:
More
Specifications
Cat. No.CA89405-790Storage temperature–2-30°C