MYP Agar Base, CRITERION™, Hardy Diagnostics
CRITERION™ MYP Agar Base for the cultivation and enumeration of Bacillus cereus from foods
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Antimicrobic Vial P, product 89404-944 and Egg Yolk Emulsion, product #233471, is required for this medium. Dehydratedculture media with engineered container.
: BSE Free
- Storage temperature:–2-30°C
Frequently Bought Together