mTEC Agar, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
À propos de cet article
CRITERION™ mTEC Agar is recommended for the differentiation and enumeration of thermotolerant Escherichia coli in water by membrane filtration and an in situ urease test
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
: BSE Free
4 Options Available Below
Product Details & Documents
CRITERION™ mTEC Agar is recommended for the differentiation and enumeration of thermotolerant Escherichia coli in water by membrane filtration and an in situ urease test
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
: BSE Free
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- CA89406-512
- CA89406-514
- CA89406-516
- CA89406-518
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Spécifications
Product Family Options
Informations sur le produit
- TailleFinal VolumeType de conditionnementAvailabilityPrice
- Supplier Number: C7740Spécifications:
More
Specifications
Cat. No.CA89406-512Storage temperature–2-30°C - Supplier Number: C7741Spécifications:
More
Specifications
Cat. No.CA89406-514Storage temperature–2-30°C - Supplier Number: C7742Spécifications:
More
Specifications
Cat. No.CA89406-516Storage temperature–2-30°C - Supplier Number: C7743Spécifications:
More
Specifications
Cat. No.CA89406-518Storage temperature–2-30°C