Modified EC Medium, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
À propos de cet article
CRITERION™ Modified EC Medium is for the detection of Escherichia coli 0157:H
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
1 Options Available Below
Product Details & Documents
CRITERION™ Modified EC Medium is for the detection of Escherichia coli 0157:H
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- CA89406-678
- CA89406-680
- CA89406-682
- CA89406-684
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Spécifications
Product Family Options
Informations sur le produit
- TailleFinal VolumeType de conditionnementAvailabilityPrice
- 122.4 g2 LZip-pouch to make 2 L, Mylar®Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CA89406-678Storage temperature–2-30°C - Supplier Number: C8091Spécifications:
More
Specifications
Cat. No.CA89406-680Storage temperature–2-30°C - 2 kgBucketDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CA89406-682Storage temperature–2-30°C - 10 kgBucketDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CA89406-684Storage temperature–2-30°C