Modified Casman, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
About this item
CRITERION™ Modified Casman for the cultivation of Haemophilus spp
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
and with added sheep blood can be used for the differentiation of Haemophilus spp. Dehydratedculture media with engineered container.
Certifications: BSE Free
4 Options Available Below
Product Details & Documents
CRITERION™ Modified Casman for the cultivation of Haemophilus spp
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
and with added sheep blood can be used for the differentiation of Haemophilus spp. Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- CA89406-444
- CA89406-446
- CA89406-448
- CA89406-450
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Specifications
Product Family Options
Product Information
- TailleFinal VolumeType de conditionnementAvailabilityPrice
- Supplier Number: C7620Specifications:
More
Specifications
Cat. No.CA89406-444Storage temperature–2-30°C - Supplier Number: C7621Specifications:
More
Specifications
Cat. No.CA89406-446Storage temperature–2-30°C - Supplier Number: C7622Specifications:
More
Specifications
Cat. No.CA89406-448Storage temperature–2-30°C - Supplier Number: C7623Specifications:
More
Specifications
Cat. No.CA89406-450Storage temperature–2-30°C