MIO Medium, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
À propos de cet article
CRITERION™ MIO Medium is recommended for use in testing motility, indole production, and ornithine decarboxylase activity of enteric bacilli
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
4 Options Available Below
Product Details & Documents
CRITERION™ MIO Medium is recommended for use in testing motility, indole production, and ornithine decarboxylase activity of enteric bacilli
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- CA89405-726
- CA89405-728
- CA89405-730
- CA89405-732
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Spécifications
Product Family Options
Informations sur le produit
- TailleFinal VolumeType de conditionnementAvailabilityPrice
- Supplier Number: C6330Spécifications:
More
Specifications
Cat. No.CA89405-726Storage temperature–2-30°C - Supplier Number: C6331Spécifications:
More
Specifications
Cat. No.CA89405-728Storage temperature–2-30°C - Supplier Number: C6332Spécifications:
More
Specifications
Cat. No.CA89405-730Storage temperature–2-30°C - Supplier Number: C6333Spécifications:
More
Specifications
Cat. No.CA89405-732Storage temperature–2-30°C