Middlebrook 7H9 Broth Base, CRITERION™, Hardy Diagnostics
Catalogue # CA89405-708
Fournisseur: HARDY DIAGNOSTICS CA
Numéro CAS:
Spécifications
- Pack type:Bottle, wide mouth
- Size:500 g
- Storage temperature:–2-30°C
- Cat. no.:CA89405-708
- Supplier No.:C6301
Spécifications
À propos de cet article
CRITERION™ Middlebrook 7H9 Broth Base is primarily for the cultivation of mycobacteria
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free