Lactose Broth, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
À propos de cet article
CRITERION™ Lactose Broth is used to detect coliforms in food, dairy products, and water, and to preenrich samples for the recovery of Salmonella species
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
4 Options Available Below
Product Details & Documents
CRITERION™ Lactose Broth is used to detect coliforms in food, dairy products, and water, and to preenrich samples for the recovery of Salmonella species
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- CA89405-528
- CA89405-530
- CA89405-532
- CA89405-534
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Spécifications
Product Family Options
Informations sur le produit
- TailleFinal VolumeType de conditionnementAvailabilityPrice
- Supplier Number: C5940Spécifications:
More
Specifications
Cat. No.CA89405-528Storage temperature–2-30°C - Supplier Number: C5941Spécifications:
More
Specifications
Cat. No.CA89405-530Storage temperature–2-30°C - Supplier Number: C5942Spécifications:
More
Specifications
Cat. No.CA89405-532Storage temperature–2-30°C - Supplier Number: C5943Spécifications:
More
Specifications
Cat. No.CA89405-534Storage temperature–2-30°C