Dermatophyte Test Medium, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
À propos de cet article
CRITERION™ Dermatophyte Test Medium is primarily for use in the isolation and detection of pathogenic dermatophytic fungi
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
: BSE Free
4 Options Available Below
Product Details & Documents
CRITERION™ Dermatophyte Test Medium is primarily for use in the isolation and detection of pathogenic dermatophytic fungi
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
: BSE Free
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- CA89405-362
- CA89405-364
- CA89405-366
- CA89405-368
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Spécifications
Product Family Options
Informations sur le produit
- TailleFinal VolumeType de conditionnementAvailabilityPrice
- Supplier Number: C5570Spécifications:
More
Specifications
Cat. No.CA89405-362Storage temperature–2-30°C - Supplier Number: C5571Spécifications:
More
Specifications
Cat. No.CA89405-364Storage temperature–2-30°C - Supplier Number: C5572Spécifications:
More
Specifications
Cat. No.CA89405-366Storage temperature–2-30°C - Supplier Number: C5573Spécifications:
More
Specifications
Cat. No.CA89405-368Storage temperature–2-30°C