Buffered Listeria Enrichment Broth (BLEB) Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Certificates
About this item
CRITERION™ Buffered Listeria Enrichment Broth (BLEB) Base is an enrichment broth for the cultivation of Listeria spp
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
from food according to the U.S. Food and Drug Administration (FDA). Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications
- Storage temperature:–2-30°C