BCSA Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
À propos de cet article
CRITERION™ BCSA Base, used for the selective isolation of Burkholderia (Pseudomonas) cepacia
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Antibiotics Polymixin B, Gentamicin, and Vancomycin required. See product label for details. Dehydratedculture media with engineered container.
: BSE Free
4 Options Available Below
Product Details & Documents
CRITERION™ BCSA Base, used for the selective isolation of Burkholderia (Pseudomonas) cepacia
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Antibiotics Polymixin B, Gentamicin, and Vancomycin required. See product label for details. Dehydratedculture media with engineered container.
: BSE Free
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- CA89406-812
- CA89406-814
- CA89406-816
- CA89406-818
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Spécifications
Product Family Options
Informations sur le produit
- TailleFinal VolumeType de conditionnementAvailabilityPrice
- Supplier Number: C8880Spécifications:
More
Specifications
Cat. No.CA89406-812Storage temperature–2-30°C - Supplier Number: C8881Spécifications:
More
Specifications
Cat. No.CA89406-814Storage temperature–2-30°C - Supplier Number: C8882Spécifications:
More
Specifications
Cat. No.CA89406-816Storage temperature–2-30°C - Supplier Number: C8883Spécifications:
More
Specifications
Cat. No.CA89406-818Storage temperature–2-30°C