Bacillus cereus Medium, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Certificates
About this item
CRITERION™ Bacillus Cereus Agar Base is a selective agar medium used for the isolation and determination of Bacillus Cereus
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Egg Yolk Emulsion, product r450290, and Bacillus Cereus Selective Supplement, Polymyxin B, product #SR099E, are required for this medium. Requires supplements: Egg Yolk Suspension 89407-700, and Bacillus Cereus Selective Supplement Polymyxin B SR099E. Dehydratedculture media with engineered container.
Certifications: BSE Free
Specifications
- Storage temperature:–2-30°C