CRITERION™ A-1 Medium, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
À propos de cet article
CRITERION™ A-1 Medium is recommended for the detection of fecal coliforms in foods, treated wastewater, and seawater.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Certifications: BSE Free
4 Options Available Below
Product Details & Documents
CRITERION™ A-1 Medium is recommended for the detection of fecal coliforms in foods, treated wastewater, and seawater.
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Certifications: BSE Free
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- CA89406-404
- CA89406-406
- CA89406-408
- CA89406-410
- Storage temperature:
- –2-30°C
- –2-30°C
- –2-30°C
- –2-30°C
Spécifications
Product Family Options
Informations sur le produit
- TailleFinal VolumeType de conditionnementAvailabilityPrice
- Supplier Number: C7570Spécifications:
More
Specifications
Cat. No.CA89406-404Storage temperature–2-30°C - Supplier Number: C7571Spécifications:
More
Specifications
Cat. No.CA89406-406Storage temperature–2-30°C - Supplier Number: C7572Spécifications:
More
Specifications
Cat. No.CA89406-408Storage temperature–2-30°C - Supplier Number: C7573Spécifications:
More
Specifications
Cat. No.CA89406-410Storage temperature–2-30°C