Calibration Baths, PolyScience
Supplier:   Polyscience
Calibration Baths, PolyScience
Supplier:  Polyscience
Certificates
Calibration Baths, PolyScience
1 Options Available Below
Product Details & Documents
PolyScience Calibration Baths accommodate up to 6 temperature measuring devices and come with 2-point factory calibration, as well as bath lids with sleeves for two 3mm, 4mm and 6mm diameter openings
Calibration Baths are available in Heated and Refrigerated/Heated models. Swivel 180™, LidDock™ and DuraTop™ features all come standard. WhisperCool® Environmental Control System is standard on refrigerated units. All models include USB-A, RS-232, RS-485 and Ethernet, and include an external temperature probe port.
Event scheduling (time and date), along with real-time clock and temperature trends (for up to 10 days) included. Drain ports make it easy to remove liquids from the bath and the washable air filter helps keep the circulator energy-efficient and running at peak performance.
Available with the Performance Programmable (OpenMode programming for unlimited steps and programs, for specific test protocols) or Performance Digital Controller, all Calibration Baths feature an icon-driven SmartTouch interface, which leads you through menus, functions and options, enabling you to make any adjustments, even with your gloves on. All screens and menus, including on-screen help, can be displayed in any of 11 different languages.
 : Complies with DIN 12876-1 Class III safety requirements for use with flammable liquids, cETLus compliant.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89400-940
  • 89400-942
  • 89400-936
  • 89400-938
  • 89400-944
  • Temperature stability:
  • ±0.005°C
  • ±0.005°C
  • ±0.005°C
  • ±0.005°C
  • ±0.005°C
  • Reservoir capacity:
  • 15L
  • 15L
  • 15L
  • 15L
  • 15L
  • Connections:
  • USB-A, Ethernet, RS-232 / RS-485, external temperature probe
  • USB-A, Ethernet, RS-232 / RS-485, external temperature probe
  • USB-A, Ethernet, RS-232 / RS-485, external temperature probe
  • USB-A, Ethernet, RS-232 / RS-485, external temperature probe
  • USB-A, Ethernet, RS-232 / RS-485, external temperature probe
Spécifications
Product Family Options
Refrigerated Callibration Baths
Informations sur le produit
  • Désignation
    Plage de température
    Electrical
    Programmable
    W×D×H
    Working Access
    Availability
    Price
  • Refrigerated/Heated Calibration Bath with Performance Programmable Controller
    –30...+200 °C
    120 V, 60 Hz
    Unlimited Steps and Programs
    54.1×22.1×77.5 cm (21.3×8.7×30.5")
    Dia.×H 11.8×28 cm (4.6×11")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $18,076.72
    $18,076.72
    Each
       $18,076.72
    Supplier Number: AP15RCAL-A11B
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    89400-944
    Temperature stability
    ±0.005°C
    Reservoir capacity
    15L
    Connections
    USB-A, Ethernet, RS-232 / RS-485, external temperature probe
    Product available on GSA Advantage®
Accessories
Informations sur le produit
  • Catalog #
    For Use With
    Details
    Availability
    Price
  • PolyScience polyclean CLARIFIER
    PolyScience polyclean CLARIFIER
    71002-500
    VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath Systems
    8 oz bottle of polyclean CLARIFIER; concentrated; treats approximately 200 gallons (757 liters)
    Log in pour la disponibilité
    Each (8oz) - $83.31
Informations sur le produit
  • Catalog #
    For Use With
    Details
    Availability
    Price
  • Teflon®-lined, Stainless Steel Overbraid Tubing
    Teflon®-lined, Stainless Steel Overbraid Tubing
    13270-868
    For Refrigerated/Heated Circulating Water Baths. Heat Only Circulating Water Baths and Open Systems. For Viscosity Baths and Calibration Baths
    Teflon®-lined Tubing, 6 mm (1/4"), Stainless Steel Overbraid with 1/4" NPT Quick Connects
    Log in pour la disponibilité
    Each (3Feet) - $322.19
Informations sur le produit
  • Catalog #
    Temperature Range
    Availability
    Price
  • Ethylene glycol, 3.8 l (1 gal.)
    Ethylene glycol, 3.8 l (1 gal.)
    71002-504
    –25...+100 °C (–13...+212 °F), when mixed 50/50 with distilled water
    Log in pour la disponibilité
    Each (1US Gal) - $253.34
Informations sur le produit
  • Catalog #
    For Use With
    Details
    Availability
    Price
  • PolyScience polyclean CLARIFIER
    PolyScience polyclean CLARIFIER
    71002-502
    VWR Refrigerated Circulating Baths, VWR Heated Circulating Baths and VWR Circulating Open Bath Systems
    case of 12 (8 oz bottles); polyclean CLARIFIER; concentrated; treats appromixately 200 gallons (757 liters)
    Log in pour la disponibilité
    Case of 12 (1/Itm/Bx) - $952.76