ContactUsLinkComponent
PROTEOSTAT® Thermal Shift Stability Assay Kit, Enzo Life Sciences
PROTEOSTAT® Thermal Shift Stability Assay Kit, Enzo Life Sciences
Catalog # 89165-918
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
PROTEOSTAT® Thermal Shift Stability Assay Kit, Enzo Life Sciences
Catalog # 89165-918
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-51027-K400
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    400 Tests
  • Storage conditions:
    –20 °C (Short Term), –80 °C (Long Term)
  • Cat. no.:
    89165-918
  • Supplier No.:
    51027-K400
Specifications
About this item
This kit has been designed for monitoring protein stability under systematic thermal stress conditions.
  • Accelerated screening for protein stability as a function of pH, ionic strength, and concentration
  • Can detect ligand binding, even without a prior knowledge of the protein’s function or ligand binding site
  • Provides a convenient, complementary orthogonal method for cross-validation of instrumentation-intensive techniques
The aggregation temperature is an indicator of protein stability and can be used to optimize conditions that minimize protein aggregation as well as to identify ligands that bind and confer structural stability to a protein of interest. Conditions that increase the aggregation temperature, increase the stability of the protein.
Delivery: Kit includes PROTEOSTAT® TS Detection Reagent, β-Lactoglobulin control, 10X Assay Buffer.