ContactUsLinkComponent
Allylamine-dUTP, Enzo Life Sciences
Allylamine-dUTP, Enzo Life Sciences
Catalog # 89165-858
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Allylamine-dUTP, Enzo Life Sciences
Catalog # 89165-858
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-42861
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Allylamine-dUTP
  • Size:
    2.5 µmoL
  • Purity detail:
    Purified by ion-exchange chromatography.
  • Purity:
    ≥93% (HPLC)
  • Concentration:
    50mM
  • Formulation:
    Liquid. Solution in water.
  • Cat. no.:
    89165-858
  • Supplier No.:
    42861
Specifications
About this item
Allylamine-2’-deoxyuridine-5’-triphosphate can replace TTP in reactions where it serves as a substrate for E. coli DNA polymerase 1 (holoenzyme and Klenow fragment), terminal deoxynucloetide transferase, T4 and Taq DNA polymerases, and reverse transcriptases (from ALV and MLV).
Allylamine-dUTP is incorporated into DNA by a variety of labeling reactions including nick translation, random primed DNA synthesis and 3’ end labeling reactions. Allylamine-labeled DNA can be efficiently conjugated to the active ester form of signal generating moieties such as fluorescent dyes, biotin and other haptens to produce labeled probes for hybridization/detection assays. The efficient incorporation of Allylamine-dUTP, in contrast to many dye-labeled nucleotides, provides for greater incorporation of signaling moieties and stronger signals.