ContactUsLinkComponent
BIO-PROBE® JC Virus Probe, Enzo Life Sciences
BIO-PROBE® JC Virus Probe, Enzo Life Sciences
Catalog # 89165-732
Supplier:  Enzo Life Sciences
CAS Number:  
BIO-PROBE® JC Virus Probe, Enzo Life Sciences
Catalog # 89165-732
Supplier:  Enzo Life Sciences
Supplier Number:  ENZ-40847
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    BIO-PROBE® JC Virus Probe
  • Size:
    2 µg
  • Concentration:
    20µg/ml
  • Formulation:
    Liquid. In 10mM TRIS HCl, pH 7.5, containing 1mM EDTA.
  • Cat. no.:
    89165-732
  • Supplier No.:
    40847
Specifications
About this item
The JC Virus BioProbe® labeled probe is prepared by nick translation of a clone of the entire JC virus genome.
JC virus, a member of the papovavirus family, usually causes subclinical infections. However, in immunocompromised individuals it can cause a fatal demyelinating brain disease, progressive multifocal leukoencephalopathy. JC virus is also neurooncogenic in primates. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).