Rat Recombinant USP2 (Catalytic Domain) (from E. coli)
Catalog # 89165-446
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Rat
- Size:100 µg
- Protein/peptide name:USP2 (catalytic domain)
- Purity:>95% (SDS-PAGE, following ion exchange chromatography)
- Concentration:0.9mg/ml
- Formulation:Liquid. In 10mM MES, containing 0.5mM DTT, 50mM TRIS-HCl, pH 7.5 and 20% glycerol.
- Cat. no.:89165-446
- Supplier No.:UW9850-0100
Specifications
About this item
Produced in E. coli. Untagged USP2 catalytic domain.