Human Recombinant Cu/Zn superoxide dismutase (from E. coli)
Catalog # 89163-974
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Size:100 µg
- Enzyme name:Cu/Zn superoxide dismutase
- Cat. no.:89163-974
- Supplier No.:SE305-0100
Specifications
About this item
Produced in E. coli.