Meloxicam ≥98%
Supplier: Enzo Life Sciences
Jump to:
À propos de cet article
COX inhibitor
Selective COX-2 inhibitor. Active in vivo. IC50 (human chondrocytes) COX-1 = 36.6µM, COX-2 = 4.7µM
Synonymes: 4-Hydroxy-2-methyl-N-(5-methylthiazol-2-yl)-2H-benzo[e][1,2]thiazine-3-carboxamide 1,1-dioxide, 4-Hydroxy-2-methyl-N-(5-methyl-2-thiazolyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
- CAS Number:71125-38-7
- Formule:C₁₄H₁₃N₃O₄S₂
- Numéro MDL:MFCD00868752
- Molecular Weight:351.40 g/mol
1 Options Available Below
Product Details & Documents
COX inhibitor
Selective COX-2 inhibitor. Active in vivo. IC50 (human chondrocytes) COX-1 = 36.6µM, COX-2 = 4.7µM
Synonymes: 4-Hydroxy-2-methyl-N-(5-methylthiazol-2-yl)-2H-benzo[e][1,2]thiazine-3-carboxamide 1,1-dioxide, 4-Hydroxy-2-methyl-N-(5-methyl-2-thiazolyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
- CAS Number:71125-38-7
- Formule:C₁₄H₁₃N₃O₄S₂
- Numéro MDL:MFCD00868752
- Molecular Weight:351.40 g/mol



