Annexin V-FITC Apoptosis Detection Kit, Enzo Life Sciences
For the detection of apoptotic cells by flow cytometry.
FITC-labelled recombinant human annexin V shows bright green fluorescence (Ex(max): 488 nm, Em(max): 530 nm).
: Kit contains Annexin V-FITC, binding buffer and propidium iodide.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Storage conditions:4 °C
- Purity:≥98% (SDS-PAGE; HPLC)
- Application:Flow Cytometry, Fluorescence Microscopy, Fluorescent Detection
Frequently Bought Together