To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Melanoma Differentiation-Associated protein 5
- Antigen symbol:MDA5
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Form:Liquid. In PBS containing 0.02% sodium azide.
- Antigen synonyms:Clinically amyopathic dermatomyositis autoantigen 140 kDa|RLR-2|Helicard|Murabutide down-regulated protein|Melanoma differentiation-associated protein 5|RNA helicase-DEAD box protein 116|MDA-5|CADM-140 autoantigen|RIG-I-like receptor 2|Helicase with 2 CARD domains|Interferon-induced with helicase C domain protein 1
- Concentration:1mg/ml
- Purification:Epitope-affinity purified
- Cat. no.:89146-318
- Supplier No.:210-935-C100
Spécifications
À propos de cet article
MDA5 is a highly conserved helicase whose ATPase activity can be stimulated by RNA and less potently by DNA. It contains two N-terminal CARD domains and a C-terminal helicase domain. The helicase domain, once freed from the CARD domains, then translocates from the cytoplasm to the nucleus where it acts on chromatin architecture. This could allow a more easy access of CAD, thereby accelerating the degradation of genomic DNA.
- For Immunoprecipitation, Western Blot
Recognizes human MDA5.
Type: Primary
Antigen: MDA5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human