ContactUsLinkComponent
p70 S6K Activity Kit, Enzo Life Sciences
Catalogue # 89141-850
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
undefined
p70 S6K Activity Kit, Enzo Life Sciences
Catalogue # 89141-850
Fournisseur:  Enzo Life Sciences
Supplier Number:  ADI-EKS-470
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    96 wells
  • Cat. no.:
    89141-850
  • Supplier No.:
    EKS-470
Spécifications
À propos de cet article
For the measurement of p70 S6K activity in partially purified, purified, or crude enzyme preparations from any species.
The p70 S6K activity kit is an efficient assay that utilizes little sample volume. This kit gives you the ability to an end-point or kinetic assay read-out in a convenient 96-well plate based assay and offers easy sample handling protocols. The assay also offers a high signal to background ratio.
Ordering information: Contents: Microtiter plate, Antibody, Conjugate, Antibody dilution buffer, Kinase assay dilution buffer, ATP, Active Kinase, Wash buffer concentrate, TMB Substrate, Stop solution 2