ContactUsLinkComponent
PKC kinase activity kit, Enzo Life Sciences
PKC kinase activity kit, Enzo Life Sciences
Catalog # 89141-838
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
PKC kinase activity kit, Enzo Life Sciences
Catalog # 89141-838
Supplier:  Enzo Life Sciences
Supplier Number:  ADI-EKS-420A
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    96 wells
  • Cat. no.:
    89141-838
  • Supplier No.:
    EKS-420A
Specifications
About this item
For the measurement of PKC kinase activity partially purified, purified, or crude enzyme preparations from any species. Cited sample type includes tissue (ref. 1).
The PKC Kinase activity kit is an efficient assay that utilizes little sample volume. This kit gives you the ability to an end-point or kinetic assay read-out in a convenient 96-well plate based assay and offers easy sample handling protocols. The assay also offers a high signal to background ratio.
Ordering information: Contents: Microtiter plate, Antibody, Conjugate, Antibody dilution buffer, Kinase assay dilution buffer, ATP, Active Kinase, Wash buffer concentrate, TMB Substrate, Stop solution 2