ContactUsLinkComponent
SOD Activity Kit, Enzo Life Sciences
Catalogue # 89141-228
Fournisseur:  Enzo Life Sciences
Numéro CAS:  
undefined
SOD Activity Kit, Enzo Life Sciences
Catalogue # 89141-228
Fournisseur:  Enzo Life Sciences
Supplier Number:  ADI-900-157
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    96 wells
  • Cat. no.:
    89141-228
  • Supplier No.:
    900-157
Spécifications
À propos de cet article
ELS offers a large collection of superoxide dismutase products for oxidative stress research.
For the measurement of total SOD activity in cell lysates and tissue from any species.
This is a versatile assay for the determination of SOD activity. Differentiate between the activity of three SOD isoenzymes: MnSOD, FeSOD, and Cu/ZnSOD. Obtain higher throughput sample analysis with a convenient colorimetric 96 well plate format and fast results in just 10 minutes.
Ordering information: Contents: Microtiter plate, Standard, SOD buffer concentrate, Xanthine oxidase, Xanthine solution, Triton x-100, WST-1 reagent