Soaker Bottle, Hach | Avantor
Soaker Bottle, Hach
Soaker Bottle
Catalog # CA97021-550
Supplier:   Hach
Fournisseur #:  5192900
Soaker Bottle, Hach
Catalog # CA97021-550
Supplier:  Hach
Soaker Bottle, Hach
Soaker Bottle, Hach
Soaker Bottle
Price
$28.04
Log in pour la disponibilité
Each
Product Details & Documents
Soaker bottle for electrode short-term and long-term storage.
Using soaker bottle for electrode storage prevents reference junction and/or sensing element from drying out to ensure best performances.
Spécifications
  • Désignation:
    Soaker Bottle