Nitrogen-Ammonia Standard Solution, 50 mg/L as NH3-N, 10 ml Voluette Ampules, Hach
CA14791-10
: Hach
:
- Concentration:50 mg/L as NH₃⁻N
- Pack type:Voluette Ampules
- Description:Nitrogen-Ammonia Standard Solution
- Size:10 ml
- Cat. no.:CA14791-10
- Supplier No.:1479110
For accuracy checks in ammonia nitrogen determinations.
- Simple Accuracy Checks
- Minimize waste
- Reliable results
: 16/pk.